2,6-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)pyridine 97%

Supplier: Ambeed
Warning

A132448-100MG A132448-250MG A132448-1G
103801-740EA 83 USD
103801-740 77136-942 77136-944 77314-320
2,6-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)pyridine 97%
2,6-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₉H₂₇N₃O₂
Boiling Pt: 467.1 °C (760 mm Hg)
Flash Pt: 236.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368372
CAS Number: 118949-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR