(±)-2,6,10,10-Tetramethyl-1-oxaspiro[4.5]-5-decene 96%

Supplier: Ambeed
Warning

Synonyms: Theaspirane

A163704-250MG A163704-1G
103805-750EA 7 USD
103805-750 103844-012 76799-108 77320-122
(±)-2,6,10,10-Tetramethyl-1-oxaspiro[4.5]-5-decene 96%
(±)-2,6,10,10-Tetramethyl-1-oxaspiro[4.5]-5-decene

Precaución: for Research Use Only

Formula: C₁₃H₂₂O
MW: 194.32 g/mol
Boiling Pt: 254.1 °C (760 mm Hg)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00085214
CAS Number: 36431-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR