Thiophene-2,5-dicarbaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,5-Thiophenedicarboxaldehyde
A507112-1G
A507112-5G
A507112-250MG
103799-666EA
12
USD
103799-666
103828-342
103839-446
76843-428
76843-430
77253-364
Thiophene-2,5-dicarbaldehyde 95%
Thiophene-2,5-dicarbaldehyde
Precaución: for Research Use Only
Formula:
C₆H₄O₂S MW: 140.16 g/mol Boiling Pt: 302 °C (760 mm Hg) Melting Pt: 115…117 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00216592 CAS Number: 932-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...