2,5-Di-tert-butyl-p-benzoquinone 98%

Supplier: Ambeed
Warning

A143577-100G A143577-500G A143577-25G A143577-5G
77247-042EA 63 USD
77247-042 77247-044 77316-576 77316-578
2,5-Di-tert-butyl-p-benzoquinone 98%
2,5-Di-tert-butyl-p-benzoquinone

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₂
Boiling Pt: 285.4 °C (760 mm Hg)
Melting Pt: 152…154 °C
Flash Pt: 106.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00019442
CAS Number: 2460-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR