N-Succinimidyl N-methylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: N-Succinimidyl methyllcarbamate, Methylcarbamic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl methylcarbamate

A112581-10G A112581-250MG A112581-1G A112581-25G
103798-046EA 7 USD
103798-046 103823-884 76718-154 76718-156 77310-694 77878-845
N-Succinimidyl N-methylcarbamate 97%
N-Succinimidyl N-methylcarbamate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₄
Melting Pt: 148…153 °C
Storage Temperature: Freezer
MDL Number: MFCD00800292
CAS Number: 18342-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR