2,5-Dimethylpyrazine 97%

Supplier: Ambeed
Warning

A129340-100G A129340-10G A129340-500G A129340-25G
77109-124EA 41.38 USD
77109-124 77109-126 77109-128 77313-650
2,5-Dimethylpyrazine 97%
2,5-Dimethylpyrazine

Precaución: for Research Use Only

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 155 °C (760 mm Hg)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00006147
CAS Number: 123-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR