2,5-Dimethylbenzoxazole 98%

Supplier: Ambeed
Warning

A100367-25G A100367-1G
103795-392EA 16 USD
103795-392 103821-092 76686-980 77296-648
2,5-Dimethylbenzoxazole 98%
2,5-Dimethylbenzoxazole

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 218.5 °C (760 mm Hg)
Melting Pt: 218…219 °C (760 mm Hg)
Flash Pt: 91.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00005773
CAS Number: 5676-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR