2,5-Dimethylbenzo[d]thiazol-6-amine 96%

Supplier: Ambeed
Warning

A284235-250MG A284235-1G
103813-630EA 194 USD
103813-630 76827-600 77305-454
2,5-Dimethylbenzo[d]thiazol-6-amine 96%
2,5-Dimethylbenzo[d]thiazol-6-amine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂S
MW: 178.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18822299
CAS Number: 686747-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR