2,5-Dimethylpyrrole 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Dimethyl-1H-pyrrole

A157607-10G A157607-1G A157607-25G A157607-5G
103805-020EA 5 USD
103805-020 103843-222 77247-540 77319-054 77319-056 77319-058
2,5-Dimethylpyrrole 98%
2,5-Dimethylpyrrole

Precaución: for Research Use Only

Formula: C₆H₉N
MW: 95.1424 g/mol
Boiling Pt: 165 °C (760 mm Hg)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00005223
CAS Number: 625-84-3
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR