2,5-Dimethylindole 98%
Supplier: Ambeed
|
Warning
|
A124657-100G
A124657-10G
A124657-25G
A124657-1G
A124657-5G
103800-566EA
6
USD
103800-566
76646-908
76646-910
76646-912
77298-014
77312-706
2,5-Dimethylindole 98%
2,5-Dimethylindole
Precaución: for Research Use Only
Formula:
C₁₀H₁₁N Melting Pt: 112…113 °C Flash Pt: 119.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005621 CAS Number: 1196-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...