2,5-Dimethoxyphenol 97%

Supplier: Ambeed
Warning

A279644-1G A279644-5G
103813-476EA 55 USD
103813-476 103853-422 77305-338 77305-340
2,5-Dimethoxyphenol 97%
2,5-Dimethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
Boiling Pt: 269.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00094237
CAS Number: 18113-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR