2,5-Dimethoxy-4-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A274614-1G A274614-25G A274614-5G
103813-204EA 29 USD
103813-204 103853-132 77092-956 77092-958 77092-960
2,5-Dimethoxy-4-methylbenzaldehyde 98%
2,5-Dimethoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD02253192
CAS Number: 4925-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR