2,5-Dihydroxyterephthalic acid 98%
Supplier: Ambeed
|
Warning
|
A173122-25G
A173122-5G
A173122-10G
A173122-500G
A173122-1G
A173122-100G
103806-794EA
8
USD
103806-794
103845-088
77082-318
77082-320
77236-318
77236-320
77301-248
77550-114
2,5-Dihydroxyterephthalic acid 98%
2,5-Dihydroxyterephthalic acid
Precaución: for Research Use Only
Formula:
C₈H₆O₆ MW: 198.13 g/mol Boiling Pt: 498.9 °C (760 mm Hg) Melting Pt: >300 °C Flash Pt: 269.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00132933 CAS Number: 610-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...