2,5-Dihydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A100800-100G A100800-10G A100800-250MG A100800-25G A100800-500G A100800-1G A100800-5G
103795-636EA 6 USD
103795-636 103821-328 77135-456 77135-458 77135-460 77135-462 77135-464 77296-842 77296-844
2,5-Dihydroxybenzaldehyde 98%
2,5-Dihydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆O₃
MW: 138.12 g/mol
Melting Pt: 97…99 °C
Flash Pt: 135.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003333
CAS Number: 1194-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR