2,5-Dihydroxy-p-benzoquinone 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dihydroxy-1,4-benzoquinone

A613393-10G A613393-1G
103840-184EA 15 USD
103840-184 77254-342 77254-344
2,5-Dihydroxy-p-benzoquinone 97%
2,5-Dihydroxy-p-benzoquinone

Precaución: for Research Use Only

Formula: C₆H₄O₄
MW: 140.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001598
CAS Number: 615-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR