2,5-Difluoropyridine 96%

Supplier: Ambeed
Danger

A449943-25G A449943-5G
103827-784EA 9 USD
103827-784 103838-854 76842-668 76842-670
2,5-Difluoropyridine 96%
2,5-Difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃F₂N
MW: 115.08
Boiling Pt: 113.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04114191
CAS Number: 84476-99-3
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR