2,5-Difluorobenzaldehyde 98%

Supplier: Ambeed
Danger

A668642-25G A668642-100G A668642-1G A668642-5G
103840-486EA 8 USD
103840-486 76844-692 77145-456 77145-458 77308-082
2,5-Difluorobenzaldehyde 98%
2,5-Difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 175.3 °C (760 mm Hg)
Flash Pt: 59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR