2,5-Difluoro-4-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A377178-1G A377178-25G A377178-5G A377178-100G A377178-10G A377178-250MG
103826-604EA 6 USD
103826-604 103837-632 76840-938 76840-940 76840-942 77003-594 77003-596 77003-598
2,5-Difluoro-4-nitrobenzoic acid 95%
2,5-Difluoro-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₂NO₄
MW: 203.10 g/mol
Boiling Pt: 364.8 °C (760 mm Hg)
Melting Pt: 145…147 °C
Flash Pt: 174.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03425705
CAS Number: 116465-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR