2,5-Difluoro-4-iodopyridine 98%
Supplier: Ambeed
|
Warning
|
A102288-10G
A102288-250MG
A102288-1G
A102288-5G
A102288-25G
A102288-100G
103796-352EA
11
USD
103796-352
103822-044
76970-214
76970-216
77308-924
77308-926
77592-078
77670-474
2,5-Difluoro-4-iodopyridine 98%
2,5-Difluoro-4-iodopyridine
Precaución: for Research Use Only
Formula:
C₅H₂F₂IN Boiling Pt: 209.7 °C (760 mm Hg) Melting Pt: 80…85 °C Flash Pt: 80.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11040606 CAS Number: 1017793-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...