2,5-Difluoro-3-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A101003-10G A101003-250MG A101003-1G
103795-746EA 28 USD
103795-746 103821-438 76780-438 76780-440 77296-944
2,5-Difluoro-3-methylbenzonitrile 98%
2,5-Difluoro-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MW: 153.129 g/mol
Boiling Pt: 211.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09996954
CAS Number: 1003712-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR