2,5-Dichloro-3-thenoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichlorothiophene-3-carboxylic acid

A123392-10G A123392-1G A123392-5G A123392-25G
103800-398EA 6 USD
103800-398 76983-340 77312-516 77312-518 77683-104
2,5-Dichloro-3-thenoic acid 97%
2,5-Dichloro-3-thenoic acid

Precaución: For research use only.

Formula: C₅H₂Cl₂O₂S
MW: 197.04 g/mol
Boiling Pt: 296.6 °C (760 mm Hg)
Melting Pt: 147…151 °C at N/A mmHg
Flash Pt: 133.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00082908
CAS Number: 36157-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR