2,5-Dichlorothiophene-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A374719-250MG A374719-5G
103826-558EA 171 USD
103826-558 103837-576 77251-820 77306-862
2,5-Dichlorothiophene-3-carbaldehyde 98%
2,5-Dichlorothiophene-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₂Cl₂OS
Storage Temperature: Refrigerator
MDL Number: MFCD01571097
CAS Number: 61200-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR