2,5-Dichloroquinoxaline 97%

Supplier: Ambeed
Warning

A118144-100MG A118144-250MG A118144-1G A118144-5G
103799-000EA 42 USD
103799-000 103824-892 76870-108 76870-110 77311-680 77311-682
2,5-Dichloroquinoxaline 97%
2,5-Dichloroquinoxaline

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
Flash Pt: 157.098 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11846477
CAS Number: 55687-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR