4-Amino-2,5-dichloropyrimidine 95%

Supplier: Ambeed
Warning

A125074-250MG A125074-1G A125074-25G
103800-622EA 19 USD
103800-622 76996-060 77312-770 77881-007
4-Amino-2,5-dichloropyrimidine 95%
4-Amino-2,5-dichloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₃Cl₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD13191633
CAS Number: 89180-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR