2,5-Dichloro-p-xylene 98%

Supplier: Ambeed
Warning

A200850-10G A200850-5G
103847-878EA 5 USD
103847-878 77112-168 77302-722
2,5-Dichloro-p-xylene 98%
2,5-Dichloro-p-xylene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 222.1 °C (760 mm Hg)
Melting Pt: 69…70 °C
Flash Pt: 86.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000610
CAS Number: 1124-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR