2,5-Dichloronicotinonitrile 97%

Supplier: Ambeed
Warning

A223127-250MG A223127-5G A223127-10G
103819-022EA 36 USD
103819-022 103849-902 76975-896 77088-518 77600-782
2,5-Dichloronicotinonitrile 97%
2,5-Dichloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₂
Boiling Pt: 255.4 °C (760 mm Hg)
Melting Pt: 118…119 °C
Flash Pt: 108.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13185481
CAS Number: 126954-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR