2,5-Dichloro-4-formylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichloroisonicotinaldehyde

A215494-100MG A215494-10G A215494-250MG A215494-1G A215494-5G
103818-496EA 17 USD
103818-496 103849-204 77030-644 77030-646 77030-648 77087-742 77087-744
2,5-Dichloro-4-formylpyridine 97%
2,5-Dichloro-4-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176.0000 g/mol
Boiling Pt: 256.3 °C (760 mm Hg)
Melting Pt: 73…74 °C
Flash Pt: 108.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06410679
CAS Number: 102645-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR