1,4-Dichloro-2-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichlorofluorobenzene

A334904-25G A334904-5G A334904-100G A334904-10G A334904-500G A334904-1G
103825-670EA 5 USD
103825-670 103836-640 76839-642 76839-644 77106-550 77106-552 77106-554 77306-476
1,4-Dichloro-2-fluorobenzene 98%
1,4-Dichloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃Cl₂F
MW: 164.99 g/mol
Boiling Pt: 70 °C at 22 mmHg
Melting Pt: 4 °C at N/A mmHg
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00060656
CAS Number: 348-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR