2,5-Dichlorobenzoic acid 98%

Supplier: Ambeed
Warning

A174610-1000G A174610-25G A174610-5G A174610-100G A174610-10G A174610-500G
103809-090EA 119 USD
103809-090 103845-258 77082-560 77082-562 77082-564 77111-134 77111-136 77111-138
2,5-Dichlorobenzoic acid 98%
2,5-Dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
Boiling Pt: 301 °C (760 mm Hg)
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002416
CAS Number: 50-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR