2,5-Dichloro-p-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichlorobenzene-1,4-diamine, 1,4-Diamino-2,5-dichlorobenzene, 2,5-Dichloro-1,4-phenylenediamine

A435045-1G A435045-25G A435045-5G A435045-100G A435045-10G
103838-656EA 21 USD
103838-656 76818-176 76842-372 76842-374 77884-120 77884-121
2,5-Dichloro-p-phenylenediamine 97%
2,5-Dichloro-p-phenylenediamine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Boiling Pt: 303.7 °C (760 mm Hg)
Melting Pt: 164…168 °C
Flash Pt: 137.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007902
CAS Number: 20103-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR