2,5-Dichlorobenzamide 97%

Supplier: Ambeed
Warning

A249064-10G A249064-250MG A249064-1G A249064-5G A249064-25G
103811-854EA 10 USD
103811-854 103851-690 76874-570 76874-572 77091-030 77091-032 77304-474
2,5-Dichlorobenzamide 97%
2,5-Dichlorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO
Boiling Pt: 262.8 °C (760 mm Hg)
Melting Pt: 170…172 °C
Flash Pt: 112.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00017609
CAS Number: 5980-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR