2,5-Dichloro-4-methylpyrimidine 97%
Supplier: Ambeed
|
Warning
|
A120109-250MG
A120109-25G
A120109-1G
A120109-5G
103799-884EA
37
USD
103799-884
77026-334
77026-336
77311-992
77311-994
2,5-Dichloro-4-methylpyrimidine 97%
2,5-Dichloro-4-methylpyrimidine
Precaución: for Research Use Only
Formula:
C₅H₄Cl₂N₂ MW: 163.0100 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13193656 CAS Number: 1192064-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...