2,5-Dichloro-4-iodopyridine 98%

Supplier: Ambeed
Warning

A133886-250MG A133886-1G
103801-926EA 11 USD
103801-926 76782-508 77298-662
2,5-Dichloro-4-iodopyridine 98%
2,5-Dichloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂IN
MW: 273.8900 g/mol
Boiling Pt: 278.7 °C (760 mm Hg)
Melting Pt: 140 °C
Flash Pt: 122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13185538
CAS Number: 796851-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR