2,5-Dichloro-4-aminophenol 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-2,5-dichlorophenol
A124574-10G
A124574-250MG
A124574-25G
A124574-1G
A124574-5G
103800-552EA
30
USD
103800-552
76971-768
76971-770
76971-772
77312-688
77312-690
2,5-Dichloro-4-aminophenol 96%
2,5-Dichloro-4-aminophenol
Precaución: for Research Use Only
Formula:
C₆H₅Cl₂NO MW: 178.0200 g/mol Boiling Pt: 295.7 °C (760 mm Hg) Melting Pt: 178…179 °C Flash Pt: 133 °C Storage Temperature: Ambient |
MDL Number:
MFCD09743861 CAS Number: 50392-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...