2,5-Dichloro-4,6-dimethylpyrimidine 95%

Supplier: Ambeed
Warning

A245851-100MG A245851-250MG
103811-674EA 124 USD
103811-674 103851-502 77013-882 77013-884
2,5-Dichloro-4,6-dimethylpyrimidine 95%
2,5-Dichloro-4,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25960625
CAS Number: 19573-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR