2,5-Dichloro-3-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A129914-1G A129914-5G A129914-250MG
103801-348EA 51 USD
103801-348 77313-754 77313-756 77670-838
2,5-Dichloro-3-(trifluoromethyl)pyridine 97%
2,5-Dichloro-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₃N
MW: 215.9880 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042244
CAS Number: 70158-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR