2,5-Dichloro-2,5-dimethylhexane 98%

Supplier: Ambeed
Warning

A243667-25G A243667-5G A243667-10G A243667-100G
103851-358EA 6 USD
103851-358 77090-564 77090-566 77249-832 77568-366
2,5-Dichloro-2,5-dimethylhexane 98%
2,5-Dichloro-2,5-dimethylhexane

Precaución: For research use only.

Formula: C₈H₁₆Cl₂
Boiling Pt: 194 °C (760 mm Hg)
Melting Pt: 63…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00126854
CAS Number: 6223-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR