2,5-Dibromothiophene-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,5-Dibromo-3-thenoic Acid
A166411-100MG
A166411-10G
A166411-250MG
A166411-25G
A166411-1G
A166411-5G
103806-064EA
11
USD
103806-064
103844-326
76973-816
76973-818
76973-820
76973-822
77320-576
77320-578
2,5-Dibromothiophene-3-carboxylic acid 97%
2,5-Dibromothiophene-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₂Br₂O₂S MW: 285.94 g/mol Boiling Pt: 345.7 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08445656 CAS Number: 7311-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...