2,5-Dibromothieno[3,2-b]thiophene 97%

Supplier: Ambeed
Danger

A799759-1G A799759-5G A799759-100MG A799759-25G
103841-058EA 131 USD
103841-058 76845-302 76845-304 77036-380 77036-382
2,5-Dibromothieno[3,2-b]thiophene 97%
2,5-Dibromothieno[3,2-b]thiophene

Precaución: for Research Use Only

Formula: C₆H₂Br₂S₂
Boiling Pt: 346.7 °C (760 mm Hg)
Melting Pt: 130 °C
Storage Temperature: Freezer
MDL Number: MFCD03931293
CAS Number: 25121-87-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR