2,5-Dibromoterephthalaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromobenzene-1,4-dicarbaldehyde, 2,5-Dibromo-1,4-diformylbenzene, 2,5-Dibromo-1,4-benzenedicarboxaldehyde

A188095-100MG A188095-10G A188095-250MG A188095-1G A188095-5G
103816-072EA 21 USD
103816-072 103846-662 77029-726 77029-728 77029-730 77084-386 77084-388
2,5-Dibromoterephthalaldehyde 97%
2,5-Dibromoterephthalaldehyde

Precaución: for Research Use Only

Formula: C₈H₄Br₂O₂
MW: 291.93 g/mol
Boiling Pt: 357.396 °C (760 mm Hg)
Melting Pt: 193…197 °C
Flash Pt: 139.818 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11521314
CAS Number: 63525-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR