2,5-Dibromopyrazine 98%
Supplier: Ambeed
|
Warning
|
A471258-10G
A471258-1G
A471258-25G
A471258-5G
A471258-100G
A471258-500G
103828-028EA
11
USD
103828-028
103839-102
77102-718
77307-478
77307-480
77307-482
77901-407
77901-777
2,5-Dibromopyrazine 98%
2,5-Dibromopyrazine
Precaución: for Research Use Only
Formula:
C₄H₂Br₂N₂ Boiling Pt: 234 °C (760 mm Hg) Flash Pt: 95.347 °C Storage Temperature: Ambient |
MDL Number:
MFCD08061601 CAS Number: 23229-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...