2,5-Dibromonicotinic acid 97%
Supplier: Ambeed
|
Warning
|
A354726-1G
A354726-5G
A354726-10G
A354726-250MG
103826-128EA
18
USD
103826-128
103837-132
76840-258
76840-260
77251-588
77251-590
2,5-Dibromonicotinic acid 97%
2,5-Dibromonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₃Br₂NO₂ MW: 280.9000 g/mol Boiling Pt: 367.1 °C (760 mm Hg) Melting Pt: 184 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06254512 CAS Number: 29312-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...