2,5-Dibromoisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromopyridine-4-carboxylic acid

A135506-100G A135506-1G A135506-25G A135506-5G
103802-162EA 27 USD
103802-162 77027-288 77298-782 77298-784 77298-786
2,5-Dibromoisonicotinic acid 98%
2,5-Dibromoisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
MW: 280.9000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08741485
CAS Number: 942473-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR