2,5-Dibromohydroquinone 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromobenzene-1,4-diol

A173172-25G A173172-5G A173172-100G A173172-10G A173172-500G A173172-1G
103806-798EA 5 USD
103806-798 103845-092 77082-328 77082-330 77248-092 77248-094 77248-096 77301-250
2,5-Dibromohydroquinone 98%
2,5-Dibromohydroquinone

Precaución: for Research Use Only

Formula: C₆H₄Br₂O₂
MW: 267.9 g/mol
Boiling Pt: 283.4 °C (760 mm Hg)
Melting Pt: 191…194 °C
Flash Pt: 125.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00192664
CAS Number: 14753-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR