2,5-Dibromoanisole 98%

Supplier: Ambeed
Warning

A250714-100G A250714-25G A250714-5G A250714-1G
103811-946EA 5 USD
103811-946 103851-784 77031-518 77091-144 77091-146 77304-538
2,5-Dibromoanisole 98%
2,5-Dibromoanisole

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
Boiling Pt: 252 °C (760 mm Hg)
Flash Pt: 99.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07780698
CAS Number: 95970-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR