3,6-Dibromo-2-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dibromo-2-methylpyridine, 2,5-Dibromo-6-methylpyridine

A197253-25G A197253-5G A197253-1000G A197253-100G A197253-10G A197253-1G A197253-500G
103847-524EA 6 USD
103847-524 77085-510 77085-512 77097-276 77097-278 77097-280 77097-282 77097-284
3,6-Dibromo-2-picoline 97%
3,6-Dibromo-2-picoline

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.92
Boiling Pt: 243.9 °C (760 mm Hg)
Melting Pt: 34…38 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06254589
CAS Number: 39919-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR