2,5-Dibromo-4-nitro-1H-imidazole 97%

Supplier: Ambeed
Warning

A156401-10G A156401-1G A156401-5G
103804-872EA 20 USD
103804-872 103843-072 77247-492 77318-844 77318-846
2,5-Dibromo-4-nitro-1H-imidazole 97%
2,5-Dibromo-4-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₃HBr₂N₃O₂
MW: 270.86 g/mol
Boiling Pt: 396 °C (760 mm Hg)
Flash Pt: 193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11877989
CAS Number: 6154-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR