2,5-Dibromo-4-methylimidazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,5-Dibromo-4-methyl-1H-imidazole
A334935-1G
A334935-250MG
103825-676EA
10
USD
103825-676
103836-646
76839-654
77114-960
2,5-Dibromo-4-methylimidazole 97%
2,5-Dibromo-4-methylimidazole
Precaución: for Research Use Only
Formula:
C₄H₄Br₂N₂ MW: 239.9 g/mol Boiling Pt: 343.1 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD02179519 CAS Number: 219814-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...