2,5-Dibromo-4-fluoroaniline 98%

Supplier: Ambeed
Warning

A341065-1G A341065-5G A341065-10G A341065-250MG
103825-824EA 19 USD
103825-824 103836-802 76839-822 76839-824 76989-450 76989-452
2,5-Dibromo-4-fluoroaniline 98%
2,5-Dibromo-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂FN
Storage Temperature: Refrigerator
MDL Number: MFCD18089317
CAS Number: 172377-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR