2,5-Dibromo-3-picoline 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Dibromo-3-methylpyridine

A272961-25G A272961-10G
103853-032EA 6 USD
103853-032 77092-822 77099-044 77305-116
2,5-Dibromo-3-picoline 98%
2,5-Dibromo-3-picoline

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.92
Boiling Pt: 260 °C (760 mm Hg)
Melting Pt: 43…47 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093085
CAS Number: 3430-18-0
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR